Assay Details
| Gene Symbol: PMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075239.2 | NP_001068707.1 | 1 - 2 | 129 | 126 | |
| GenBank mRNA | BC120137.1 | - | 1 - 2 | 130 | 126 | |
More Information
Set Membership: |
|

PMP2
Genomic Map