Assay Details
| Gene Symbol: TMEM59L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075301.1 | NP_001068769.1 | 4 - 5 | 642 | 62 | |
| GenBank mRNA | BC120018.1 | - | 4 - 5 | 642 | 62 | |
More Information
Set Membership: |
|

TMEM59L
Genomic Map