Assay Details
| Gene Symbol: CYP4F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075322.1 | NP_001068790.1 | 11 - 12 | 1753 | 60 | |
| GenBank mRNA | BT026323.1 | - | 11 - 12 | 1753 | 60 | |
More Information
Set Membership: |
|

CYP4F2
Genomic Map