Assay Details
| Gene Symbol: VMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075368.2 | NP_001068836.1 | 1 - 2 | 79 | 112 | |
| GenBank mRNA | BC120116.1 | - | 1 - 2 | 80 | 112 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120116.1, NM_001075368.2 transcripts.
Set Membership: |
|

VMP1
Genomic Map