Assay Details
| Gene Symbol: VMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075368.2 | NP_001068836.1 | 7 - 8 | 821 | 129 | |
| GenBank mRNA | BC120116.1 | - | 7 - 8 | 822 | 129 | |
More Information
Set Membership: |
|

VMP1
Genomic Map