Assay Details
| Gene Symbol: TMEM45A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075405.1 | NP_001068873.1 | 2 - 3 | 488 | 82 | |
| GenBank mRNA | BC116005.1 | - | 2 - 3 | 506 | 82 | |
More Information
Set Membership: |
|

TMEM45A
Genomic Map