Assay Details
| Gene Symbol: TRGC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC120033.1 | - | 5 - 6 | 840 | 110 | |
| X63680.1 | - | 535 | 110 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRGC4| Gene Symbol: TRGC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC120033.1 | - | 5 - 6 | 840 | 110 | |
| X63680.1 | - | 535 | 110 | |||
Set Membership: |
|