Assay Details
| Gene Symbol: PSKH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075413.1 | NP_001068881.1 | 1 - 2 | 117 | 120 | |
| GenBank mRNA | BT026549.1 | - | 1 - 2 | 117 | 120 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001075413.1 transcripts.
Set Membership: |
|

PSKH1
Genomic Map