Assay Details
| Gene Symbol: DECR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075423.1 | NP_001068891.1 | 5 - 6 | 566 | 83 | |
| GenBank mRNA | BC122606.1 | - | 5 - 6 | 566 | 83 | |
More Information
Set Membership: |
|

DECR1
Genomic Map