Assay Details
| Gene Symbol: STARD3NL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075427.2 | NP_001068895.1 | 8 - 9 | 842 | 119 | |
| GenBank mRNA | BC120111.1 | - | 8 - 9 | 843 | 119 | |
More Information
Set Membership: |
|

STARD3NL
Genomic Map