Assay Details
| Gene Symbol: CMTM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075434.1 | NP_001068902.1 | 1 - 2 | 169 | 71 | |
| GenBank mRNA | BC118492.1 | - | 1 - 2 | 169 | 71 | |
More Information
Set Membership: |
|

CMTM5
Genomic Map