Assay Details
| Gene Symbol: SETMAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075508.1 | NP_001068976.1 | 1 - 2 | 177 | 94 | |
| GenBank mRNA | BC119874.1 | - | 1 - 2 | 177 | 94 | |
More Information
Set Membership: |
|

SETMAR
Genomic Map