Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075592.1 | NP_001069060.1 | 2 - 3 | 430 | 74 | |
| GenBank mRNA | BC120333.1 | - | 2 - 3 | 430 | 74 | |
More Information
Set Membership: |
|

MANSC1
Genomic Map