Assay Details
| Gene Symbol: RGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075596.1 | NP_001069064.1 | 1 | 238 | 61 | |
| GenBank mRNA | BC120026.1 | - | 1 | 238 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGS2| Gene Symbol: RGS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075596.1 | NP_001069064.1 | 1 | 238 | 61 | |
| GenBank mRNA | BC120026.1 | - | 1 | 238 | 61 | |
Set Membership: |
|