Assay Details
| Gene Symbol: PDCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075645.1 | NP_001069113.1 | 5 - 6 | 638 | 66 | |
| GenBank mRNA | BC119960.1 | - | 5 - 6 | 638 | 66 | |
More Information
Set Membership: |
|

PDCL3
Genomic Map