Assay Details
| Gene Symbol: TP53I11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075725.1 | NP_001069193.1 | 5 - 6 | 489 | 69 | |
| GenBank mRNA | BC120050.1 | - | 5 - 6 | 489 | 69 | |
More Information
Set Membership: |
|

TP53I11
Genomic Map