Assay Details
| Gene Symbol: SEC14L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075765.1 | NP_001069233.1 | 11 - 12 | 1130 | 55 | |
| GenBank mRNA | BC120194.1 | - | 11 - 12 | 1130 | 55 | |
More Information
Set Membership: |
|

SEC14L4
Genomic Map