Assay Details
| Gene Symbol: SEC14L4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075765.1 | NP_001069233.1 | 9 - 10 | 820 | 73 | |
| GenBank mRNA | BC120194.1 | - | 9 - 10 | 820 | 73 | |
| Gene Symbol: SEC14L6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC149922.1 | - | 9 - 10 | 2030 | 73 | |
More Information
Set Membership: |
|

SEC14L4
Genomic Map