Assay Details
| Gene Symbol: AADAC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075791.2 | NP_001069259.1 | 2 - 3 | 383 | 122 | |
| GenBank mRNA | BC120261.1 | - | 2 - 3 | 384 | 122 | |
More Information
Set Membership: |
|

AADAC
Genomic Map