Assay Details
| Gene Symbol: MTFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075863.2 | NP_001069331.1 | 2 - 3 | 294 | 109 | |
| GenBank mRNA | BC120145.1 | - | 2 - 3 | 295 | 109 | |
More Information
Set Membership: |
|

MTFP1
Genomic Map