Assay Details
| Gene Symbol: MTFP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075863.2 | NP_001069331.1 | 3 - 4 | 535 | 95 | |
| GenBank mRNA | BC120145.1 | - | 3 - 4 | 536 | 95 | |
More Information
Set Membership: |
|

MTFP1
Genomic Map