Assay Details
| Gene Symbol: DOHH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075886.1 | NP_001069354.1 | 3 - 4 | 659 | 81 | |
| GenBank mRNA | BC120351.1 | - | 3 - 4 | 659 | 81 | |
| DQ990881.1 | - | 3 - 4 | 592 | 81 | ||
| DQ990882.1 | - | 3 - 4 | 592 | 81 | ||
| DQ990883.1 | - | 3 - 4 | 592 | 81 | ||
| DQ990884.1 | - | 3 - 4 | 337 | 81 | ||
More Information
Set Membership: |
|

DOHH
Genomic Map