Assay Details
| Gene Symbol: GP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075950.2 | NP_001069418.2 | 9 - 10 | 1622 | 68 | |
| GenBank mRNA | BC122737.1 | - | 8 - 9 | 1615 | 68 | |
| EV723489.1 | - | 2 - 3 | 548 | 68 | ||
More Information
Set Membership: |
|

GP2
Genomic Map