Assay Details
| Gene Symbol: DEDD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076017.1 | NP_001069485.1 | 4 - 5 | 707 | 69 | |
| GenBank mRNA | BC120032.1 | - | 4 - 5 | 707 | 69 | |
More Information
Set Membership: |
|

DEDD2
Genomic Map