Assay Details
| Gene Symbol: FAR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076022.1 | NP_001069490.1 | 2 - 3 | 379 | 61 | |
| GenBank mRNA | BC119968.1 | - | 2 - 3 | 379 | 61 | |
More Information
Set Membership: |
|

FAR2
Genomic Map