Assay Details
| Gene Symbol: EFEMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076049.1 | NP_001069517.1 | 3 - 4 | 279 | 79 | |
| GenBank mRNA | BC122660.1 | - | 3 - 4 | 279 | 79 | |
More Information
Set Membership: |
|

EFEMP2
Genomic Map