Assay Details
| Gene Symbol: TMLHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076064.1 | NP_001069532.1 | 7 - 8 | 1247 | 74 | |
| GenBank mRNA | BC120318.1 | - | 7 - 8 | 1247 | 74 | |
More Information
Set Membership: |
|

TMLHE
Genomic Map