Assay Details
| Gene Symbol: TTLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076171.1 | NP_001069639.1 | 3 - 4 | 465 | 59 | |
| GenBank mRNA | BC120322.1 | - | 3 - 4 | 465 | 59 | |
More Information
Set Membership: |
|

TTLL1
Genomic Map