Assay Details
| Gene Symbol: RNF168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076289.2 | NP_001069757.2 | 6 - 7 | 1085 | 83 | |
| GenBank mRNA | BC122577.1 | - | 5 - 6 | 963 | 83 | |
| DY143566.1 | - | 3 - 4 | 278 | 83 | ||
More Information
Set Membership: |
|

RNF168
Genomic Map