Assay Details
| Gene Symbol: CDK2AP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076369.2 | NP_001069837.1 | 2 - 3 | 445 | 81 | |
| GenBank mRNA | BC123623.1 | - | 2 - 3 | 446 | 81 | |
More Information
Set Membership: |
|

CDK2AP1
Genomic Map