Assay Details
| Gene Symbol: FRA10AC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076392.1 | NP_001069860.1 | 9 - 10 | 774 | 112 | |
| GenBank mRNA | BC122578.1 | - | 9 - 10 | 774 | 112 | |
More Information
Set Membership: |
|

FRA10AC1
Genomic Map