Assay Details
| Gene Symbol: SRSF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076394.1 | NP_001069862.1 | 2 - 3 | 465 | 62 | |
| GenBank mRNA | BC119930.1 | - | 2 - 3 | 465 | 62 | |
More Information
Set Membership: |
|

SRSF1
Genomic Map