Assay Details
| Gene Symbol: FN3KRP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076399.1 | NP_001069867.1 | 4 - 5 | 487 | 68 | |
| GenBank mRNA | BC120335.1 | - | 4 - 5 | 487 | 68 | |
More Information
Set Membership: |
|

FN3KRP
Genomic Map