Assay Details
| Gene Symbol: TAFA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076435.1 | NP_001069903.1 | 10 - 11 | 1065 | 66 | |
| GenBank mRNA | BC120098.1 | - | 10 - 11 | 1065 | 66 | |
More Information
Set Membership: |
|

TAFA4
Genomic Map