Assay Details
| Gene Symbol: ST14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076538.1 | NP_001070006.1 | 15 - 16 | 1914 | 58 | |
| GenBank mRNA | BC122638.1 | - | 15 - 16 | 1914 | 58 | |
More Information
Set Membership: |
|

ST14
Genomic Map