Assay Details
| Gene Symbol: CDH18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076837.1 | NP_001070305.1 | 2 - 3 | 532 | 62 | |
| GenBank mRNA | BC123714.1 | - | 2 - 3 | 532 | 62 | |
More Information
Set Membership: |
|

CDH18
Genomic Map