Assay Details
| Gene Symbol: WFIKKN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076885.1 | NP_001070353.1 | 1 - 2 | 406 | 66 | |
| GenBank mRNA | BC123391.1 | - | 1 - 2 | 406 | 66 | |
More Information
Set Membership: |
|

WFIKKN2
Genomic Map