Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076914.1 | NP_001070382.2 | 3 - 4 | 270 | 90 | |
| GenBank mRNA | BC123537.1 | - | 3 - 4 | 270 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC123537.1, NM_001076914.1 transcripts.
Set Membership: |
|

ZNF527
Genomic Map