Assay Details
| Gene Symbol: GOLGA7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076954.1 | NP_001070422.1 | 1 - 2 | 135 | 83 | |
| GenBank mRNA | BC122697.1 | - | 1 - 2 | 135 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC122697.1, NM_001076954.1 transcripts.
Set Membership: |
|

GOLGA7B
Genomic Map