Assay Details
| Gene Symbol: GOLGA7B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076954.1 | NP_001070422.1 | 3 - 4 | 401 | 68 | |
| GenBank mRNA | BC122697.1 | - | 3 - 4 | 401 | 68 | |
More Information
Set Membership: |
|

GOLGA7B
Genomic Map