Assay Details
| Gene Symbol: ERICH5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077844.1 | NP_001071312.1 | 1 - 2 | 174 | 60 | |
| GenBank mRNA | BC123516.1 | - | 1 - 2 | 174 | 60 | |
More Information
Set Membership: |
|

ERICH5
Genomic Map