Assay Details
| Gene Symbol: DNAJC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078102.1 | NP_001071570.1 | 4 - 5 | 365 | 80 | |
| GenBank mRNA | BC120442.1 | - | 4 - 5 | 365 | 80 | |
More Information
Set Membership: |
|

DNAJC24
Genomic Map