Assay Details
| Gene Symbol: GNRH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078137.1 | NP_001071605.1 | 1 - 2 | 56 | 114 | |
| GenBank mRNA | BC120472.1 | - | 1 - 2 | 56 | 114 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC120472.1, NM_001078137.1 transcripts.
Set Membership: |
|

GNRH1
Genomic Map