Assay Details
| Gene Symbol: CMC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078151.1 | NP_001071619.1 | 2 - 3 | 456 | 81 | |
| GenBank mRNA | BC120453.1 | - | 2 - 3 | 456 | 81 | |
More Information
Set Membership: |
|

CMC4
Genomic Map