Assay Details
| Gene Symbol: THAP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080741.2 | NP_001074210.1 | 1 | 1451 | 89 | |
| GenBank mRNA | BC122787.1 | - | 1 | 1452 | 89 | |
More Information
Set Membership: |
|

THAP10
Genomic Map