Assay Details
| Gene Symbol: COQ4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083672.2 | NP_001077141.1 | 2 - 3 | 242 | 61 | |
| GenBank mRNA | BC122805.1 | - | 2 - 3 | 243 | 61 | |
More Information
Set Membership: |
|

COQ4
Genomic Map