Assay Details
| Gene Symbol: ADM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173888.3 | NP_776313.1 | 4 - 5 | 503 | 73 | |
| GenBank mRNA | AB055107.1 | - | 3 - 4 | 401 | 73 | |
| AJ001613.1 | - | 3 - 4 | 400 | 73 | ||
| BC123826.1 | - | 4 - 5 | 503 | 73 | ||
More Information
Set Membership: |
|

ADM
Genomic Map