Assay Details
| Gene Symbol: CRYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174025.3 | NP_776450.2 | 9 - 10 | 1191 | 87 | |
| GenBank mRNA | BC119936.1 | - | 9 - 10 | 1191 | 87 | |
| U70048.1 | - | 8 - 9 | 883 | 87 | ||
More Information
Set Membership: |
|

CRYZ
Genomic Map