Assay Details
| Gene Symbol: YWHAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174794.2 | NP_777219.2 | 4 - 5 | 785 | 63 | |
| GenBank mRNA | AF043736.1 | - | 3 - 4 | 578 | 63 | |
| BC120112.1 | - | 4 - 5 | 785 | 63 | ||
More Information
Set Membership: |
|

YWHAB
Genomic Map