Assay Details
| Gene Symbol: APOBEC3Z2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001346123.1 | NP_001333052.1 | 1 - 2 | 141 | 102 | |
| GenBank mRNA | DQ974646.1 | - | 1 - 2 | 97 | 102 | |
| EU864535.1 | - | 1 - 2 | 144 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001346123.1 transcripts.
Set Membership: |
|

APOBEC3Z2
Genomic Map